Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFRS18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104168
Description
Antibody detects endogenous levels of total SFRS18.
SFRS18 is a protein coding gene.
Specifications
SFRS18 | |
Polyclonal | |
Unconjugated | |
PNISR | |
5730406M06Rik; AA673174; argi; arginine/serine-rich protein PNISR; bA98I9.2; C6orf111; HSPC261; HSPC306; Pinsr; PNISR; PNN interacting serine and arginine rich protein; PNN interacting serine/arginine-rich; PNN-interacting serine/arginine-rich protein; RGD1307395; RP11-98I9.2; serine/arginine-rich splicing factor 18; serine/arginine-rich-splicing regulatory protein 130; serine-arginine-rich-splicing regulatory protein 130; Sfrs18; splicing factor, arginine/serine-rich 130; Splicing factor, arginine/serine-rich 18; SR rich protein; SR-related protein; SR-rich protein; SRrp130; Srsf18 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25957, 297942, 66625 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2AJT4, Q8TF01 | |
PNISR | |
A synthesized peptide derived from human PNISR(Accession Q8TF01), corresponding to amino acid residues G177-V227. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction