Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFRS18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558544
Description
Immunogen sequence: PLNQQQWMQS FQHQQDPSQI DWAALAQAWI AQREASGQQS MVEQPPGMMP NGQDMSTMES GPNNHGNFQG DSNFNRMWQP EWGMHQQPPH PPPDQPWMPP TPGPMDIVPP SEDSNSQDSG EFAPDNRHIF NQNNHNFGG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
SFRS18 is a protein coding gene.
Specifications
SFRS18 | |
Polyclonal | |
Unconjugated | |
PNISR | |
5730406M06Rik; AA673174; argi; arginine/serine-rich protein PNISR; bA98I9.2; C6orf111; HSPC261; HSPC306; Pinsr; PNISR; PNN interacting serine and arginine rich protein; PNN interacting serine/arginine-rich; PNN-interacting serine/arginine-rich protein; RGD1307395; RP11-98I9.2; serine/arginine-rich splicing factor 18; serine/arginine-rich-splicing regulatory protein 130; serine-arginine-rich-splicing regulatory protein 130; Sfrs18; splicing factor, arginine/serine-rich 130; Splicing factor, arginine/serine-rich 18; SR rich protein; SR-related protein; SR-rich protein; SRrp130; Srsf18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF01 | |
PNISR | |
Recombinant protein corresponding to Human SFRS18. Recombinant protein control fragment (Product #RP-90224). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction