Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599713 Supplier Invitrogen™ Supplier No. PA599713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCAF1.

Splicing factor that seems to antagonize SR proteins in pre-mRNA splicing regulation. Also binds the repetitive C-terminal domain (CTD) of RNA polymerase II; may act with other CTD binding proteins to link transcription and pre-mRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SCAF1
Gene Accession No. Q5U4C3, Q9H7N4
Gene Alias AI480556; CTD-binding SR-like protein rA1; CTD-binding SR-like rA1; SCAF; Scaf1; Serine arginine-rich pre-mRNA splicing factor SR-A1; SFRS19; Splicing factor, arginine/serine-rich 19; SRA1; Sr-a1; SR-related and CTD-associated factor 1; SR-related CTD associated factor 1; SR-related CTD-associated factor 1; SR-related C-terminal domain-associated factor 1; SR-related-CTD-associated factor
Gene Symbols SCAF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCAF1(Accession Q9H7N4), corresponding to amino acid residues V43-T93.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233208, 58506
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.