Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS2IP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559008
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9008 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9008 Supplier Bethyl Laboratories, Inc Supplier No. A301007AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.

Specifications

Antigen SFRS2IP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SCAF11
Gene Accession No. Q99590
Gene Alias CASP11, CTD-associated SR protein 11, renal carcinoma antigen NY-REN-40, SC35-interacting protein 1, serine/arginine-rich splicing factor 2, interacting protein, serine/arginine-rich splicing factor 2-interacting protein, SFRS2-interacting protein, SFRS2IP, SIP1, splicing factor, arginine/serine-rich 2, interacting protein, splicing factor, arginine/serine-rich 2-interacting protein, splicing regulatory protein 129, SR-related and CTD-associated factor 11, SR-related CTD-associated factor 11, SRRP129, SRSF2-interacting protein, SRSF2IP
Gene Symbols SCAF11
Host Species Rabbit
Immunogen Between 1098 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9169
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.