Learn More
SFRS2IP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301007A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.Specifications
| SFRS2IP | |
| Polyclonal | |
| Unconjugated | |
| SCAF11 | |
| CASP11, CTD-associated SR protein 11, renal carcinoma antigen NY-REN-40, SC35-interacting protein 1, serine/arginine-rich splicing factor 2, interacting protein, serine/arginine-rich splicing factor 2-interacting protein, SFRS2-interacting protein, SFRS2IP, SIP1, splicing factor, arginine/serine-rich 2, interacting protein, splicing factor, arginine/serine-rich 2-interacting protein, splicing regulatory protein 129, SR-related and CTD-associated factor 11, SR-related CTD-associated factor 11, SRRP129, SRSF2-interacting protein, SRSF2IP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9169 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q99590 | |
| SCAF11 | |
| Between 1098 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.