Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS2IP Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 280981AP150UL

Description
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.Specifications
SFRS2IP | |
Polyclonal | |
Unconjugated | |
SCAF11 | |
CASP11, CTD associated SR protein 11, NY-REN-40, SC35 interacting protein 1, SCAF11, SFRS2IP, SIP1, SRRP129, SRSF2 interacting protein, SRSF2IP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9169 | |
-20°C | |
Liquid |
Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99590 | |
SCAF11 | |
SFRS2IP Fusion Protein Ag24381 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction