Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS3 Rabbit anti-Human, Mouse, Rat, Clone: 9A1Z4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | SFRS3 |
---|---|
Clone | 9A1Z4 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
SFRS3 Monoclonal antibody specifically detects SFRS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SFRS3 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6428 | |
IgG | |
Affinity purified |
9A1Z4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Pre-mRNA-splicing factor SRP20, serine/arginine-rich splicing factor 3, SFRS3splicing factor, arginine/serine-rich, 20-kD, Splicing factor, arginine/serine-rich 3SRP20, SRp20 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SFRS3 (P84103). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title