Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS4 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204144AF405
Description
SFRS4 Polyclonal antibody specifically detects SFRS4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SFRS4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Pre-mRNA-splicing factor SRP75, serine/arginine-rich splicing factor 4, SFRS4, Splicing factor, arginine/serine-rich 4SRP001LB, SRP75SR splicing factor 4 | |
The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human Pre-mRNA-Splicing Factor SRP75 using the numbering given in entry NP_005617.2 (Gene ID 6429). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6429 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction