Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204143
Description
SFRS4 Polyclonal specifically detects SFRS4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
SFRS4 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Pre-mRNA-splicing factor SRP75, serine/arginine-rich splicing factor 4, SFRS4, Splicing factor, arginine/serine-rich 4SRP001LB, SRP75SR splicing factor 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
6429 | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-10 mcg/mg | |
NP_005617.2 | |
SRSF4 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human Pre-mRNA-Splicing Factor SRP75 using the numbering given in entry NP_005617.2 (GeneID 6429). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction