Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984084
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4084 Supplier Bioss Supplier No. BS17300R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.

Specifications

Antigen SFRS4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SRSF4
Gene Accession No. P84103, Q08170, Q13243, Q13247
Gene Alias 1210001E11Rik; 1300011P19Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; D11Mgi27; Delayed-early protein HRS; Docking protein alpha; DP-alpha; epididymis secretory protein Li 91; HEL-S-91; HRS; Insulin-induced growth response protein CL-4; LOC100340192; LOC100347748; MNCb-2616; pre-mRNA splicing factor SRp20; Pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; Pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RCJMB04_20j14; RCJMB04_6g11; RGD1561347; serine and arginine rich splicing factor 3; serine and arginine rich splicing factor 4; serine and arginine rich splicing factor 5; serine and arginine rich splicing factor 6; Serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; Serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; Sfrs3; SFRS4; SFRS5; Sfrs6; signal recognition particle receptor (′docking protein′); Signal recognition particle receptor subunit alpha; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor SRp55; splicing factor SRp55-1; Splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); Splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); Splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); Splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SR-alpha; SRP001LB; SRP20; SRP40; Srp55; SRP55-1; SRP75; Srpr; Srpra; Srsf3; SRSF4; SRSF5; SRSF6; X16
Gene Symbols SRSF3, SRSF4, SRSF5, SRSF6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SFRS4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6428, 6429, 6430, 6431
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.