Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559050 Supplier Invitrogen™ Supplier No. PA559050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VELPPTAKMH AIIERTASFV CRQGAQFEIM LKAKQARNSQ FDFLRFDHYL NPYYKFIQKA MKEGRYTVLA ENKSDEKKKS GVSSDNEDDD D Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SFSWAP
Gene Accession No. Q12872
Gene Alias 1190005N23Rik; 6330437E22Rik; AI197402; AW212079; serine/arginine-rich splicing factor 8; Sfrs8; SFSWAP; splicing factor SWAP homolog; splicing factor, arginine/serine-rich 8; splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila); splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, Drosophila homolog); splicing factor, suppressor of white-apricot family; splicing factor, suppressor of white-apricot homolog; splicing factor, suppressor of white-apricot homolog (Drosophila); Srsf8; Suppressor of white apricot protein homolog; SWAP
Gene Symbols SFSWAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SFRS8. Recombinant protein control fragment (Product #RP-97659).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.