Learn More
SFRS8/SWAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | SFRS8/SWAP |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2182
![]() |
Bethyl Laboratories, Inc
IHC00491T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2181
![]() |
Bethyl Laboratories, Inc
IHC00491 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla and Crab-eating macaque
SWAP (suppressor of white apricot protein homolog), also known as SFRS8 (splicing factor, arginine/serine-rich 8), is the 951 amino acid human homolog of a Drosophilia splicing protein. Localized to the nucleus, SWAP contains two SURP repeats through which it is thought to mediate splicing events, possibly regulating the alternative splicing of Fibronectin and CD45RC. SWAP regulates its own expression levels (via control of splicing in its first two introns) and may act in tandem with other arginine/serine-rich splicing factors to control protein expression. The gene encoding SWAP is located on a region of chromosome 12 that is related to asthma susceptibility, possibly indicating a role for SWAP in the development of asthma. Multiple isoforms of SWAP exist due to alternative splicing events.Specifications
SFRS8/SWAP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Splicing factor, suppressor of white-apricot homolog, Splicing factor, arginine/serine-rich 8, Suppressor of white apricot protein homolog, SFSWAP, SFRS8, SWAP | |
SFSWAP | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q12872 | |
6433 | |
Between 500 and 550 | |
Primary | |
4° C | |
SFSWAP |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.