Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157465
Description
SFRS9 Polyclonal specifically detects SFRS9 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SFRS9 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Pre-mRNA-splicing factor SRp30C, serine/arginine-rich splicing factor 9, SFRS9, Splicing factor, arginine/serine-rich 9SR splicing factor 9, SRp30c | |
Rabbit | |
Affinity purified | |
RUO | |
8683 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
Q13242 | |
SRSF9 | |
Synthetic peptides corresponding to SFRS9(splicing factor, arginine/serine-rich 9) The peptide sequence was selected from the middle region of SFRS9. Peptide sequence VCYADVQKDGVGMVEYLRKEDMEYALRKLDDTKFRSHEGETSYIRVYPER. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Caenorhabditis briggsae AF16: 100%; Caenorhabditis elegans: 100%; Western clawed frog: 100%; Xenopus: 100%; Rat: 100%; Mouse: 100%; Caenorhabditis vulgaris: 100%; Canine: 100%; Human: 100%; Zebrafish: 92%; Chicken: 92%; Sumatran orangutan: 92%; Green puffer: 92%; Pig: 92%; Eye worm: 92%; Atlantic salmon: 92%; Filarial nematode worm: 92%; Bovine: 92%; Trichoplax reptans: 91%; Florida lancelet: 90%; Starlet sea anemone: 85%; Blood fluke: 85%; Pea aphid: 85%; Yellowfever mosquito: 84%; Camponotus floridanus: 84%; Body louse: 84%; Savannah tsetse fly: 84%; Red flour beetle: 84%; Black-legged tick: 84%; Fruit fly: 84%; Southern house mosquito: 84%; Midge: 84%; Harpegnathos saltator: 81%; African malaria mosquito: 81%;. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction