Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS9 Antibody (OTI5G7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274152FR
Description
SFRS9 Monoclonal specifically detects SFRS9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SFRS9 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
SRSF9 | |
Full length recombinant protein of human SFRS9 (NP_003760) produced in E.coli. | |
0.1 mL | |
8683.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5G7 | |
Western Blot, Immunohistochemistry | |
Pre-mRNA-splicing factor SRp30C, serine/arginine-rich splicing factor 9, SFRS9, Splicing factor, arginine/serine-rich 9SR splicing factor 9, SRp30c | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction