Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFT2B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2835140.1ML
Description
SFT2B Polyclonal specifically detects SFT2B in Human samples. It is validated for Western Blot.Specifications
| SFT2B | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 375035 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| dJ747L4.C1.2, FLJ34085, SFT2 domain containing 2, SFT2 domain-containing protein 2, UNQ512, vesicle transport protein SFT2B | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human SFT2B | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction