Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFT2D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599752 Supplier Invitrogen™ Supplier No. PA599752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFT2D3.

SFT2D3 may be involved in fusion of retrograde transport vesicles derived from an endocytic compartment with the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFT2D3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SFT2D3
Gene Accession No. Q587I9, Q9CSV6
Gene Alias 2610206C24Rik; AI847965; AW046173; RGD1561764; SFT2 domain containing 3; SFT2 domain-containing protein 3; Sft2d3; vesicle transport protein SFT2C
Gene Symbols SFT2D3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SFT2D3(Accession Q587I9), corresponding to amino acid residues Y11-A61.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67158, 84826
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.