Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPA1 Rabbit anti-Human, Unconjugated, Clone: RM334, Abnova™

Catalog No. 89339194 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-194 Supplier Abnova Corporation Supplier No. MAB21977
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SFTPA1 Monoclonal antibody specifically detects SFTPA1 in Human samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen SFTPA1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM334
Conjugate Unconjugated
Description surfactant protein A1
Gene SFTPA1
Gene Alias COLEC4, FLJ51913, SFTP1, SP-A, SP-A1
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human SFTPA1
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 653509
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.