Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFTPA1/SFTPA2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA538059
Description
This antibody is predicted to react with cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Surfactant protein A (SP-A) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-D and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages. The serum concentration is 45 ng/mL in healthy individuals.
Specifications
SFTPA1/SFTPA2 | |
Polyclonal | |
Unconjugated | |
Sftpa1 | |
35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC4; COLEC5; collectin 5; collectin-4; Collectin-5; FLJ50593; FLJ51913; FLJ61144; FLJ77898; FLJ79095; FLJ99559; MGC133365; MGC198590; PSAP; PSPA; PSP-A; pulmonary surfactant-associated protein A; Pulmonary surfactant-associated protein A1; pulmonary surfactant-associated protein A2; Sftp1; Sftp-1; Sftpa; Sftpa1; SFTPA1B; SFTPA2; SFTPA2B; Sftpl; SP-2A; SP-2A beta; SP-2A gamma; SPA; SP-A; SPA1; SP-A1; SP-A1 beta; SP-A1 delta; SP-A1 epsilon; SP-A1 gamma; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant associated protein A; surfactant associated protein A1; surfactant protein A1; surfactant protein A1 variant AB'D' 6A; surfactant protein A1 variant AB'D' 6A2; surfactant protein A1 variant AB'D' 6A3; surfactant protein A1 variant AB'D' 6A4; surfactant protein A1 variant ACD' 6A; surfactant protein A1 variant ACD' 6A2; surfactant protein A1 variant ACD' 6A3; surfactant protein A1 variant ACD' 6A4; surfactant protein A1 variant AD' 6A; surfactant protein A1 variant AD' 6A2; surfactant protein A1 variant AD' 6A3; surfactant protein A1 variant AD' 6A4; surfactant protein A1B; surfactant protein A2; surfactant pulmonary associated protein A1; surfactant, pulmonary-associated protein A1; surfactant, pulmonary-associated protein A1A; surfactant, pulmonary-associated protein A1B; surfactant, pulmonary-associated protein A2A; Surfactant-associated protein 1 (pulmonary surfactant protein SP-A); Surfactant-associated protein 1 (pulmonary surfactant protein, SP-A) | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
653509, 729238 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8IWL1, Q8IWL2 | |
Sftpa1, SFTPA2 | |
Peptide with sequence C-HLDEELQATLHDFR, from the internal region of SFTPA1 (aa102-115). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction