Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFTPA1/SFTPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579987
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat lung tissue, mouse lung tissue. IHC: Mouse Lung tissue, Rat Lung tissue, Human Lung Cancer tissue IHC-F: Mouse Lung tissue, Rat Lung tissue.
Surfactant protein A (SP-A) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-D and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages. The serum concentration is 45 ng/mL in healthy individuals.
Specifications
| SFTPA1/SFTPA2 | |
| Polyclonal | |
| Unconjugated | |
| Sftpa1 | |
| 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC4; COLEC5; collectin 5; collectin-4; Collectin-5; FLJ50593; FLJ51913; FLJ61144; FLJ77898; FLJ79095; FLJ99559; MGC133365; MGC198590; PSAP; PSPA; PSP-A; pulmonary surfactant-associated protein A; Pulmonary surfactant-associated protein A1; pulmonary surfactant-associated protein A2; Sftp1; Sftp-1; Sftpa; Sftpa1; SFTPA1B; SFTPA2; SFTPA2B; Sftpl; SP-2A; SP-2A beta; SP-2A gamma; SPA; SP-A; SPA1; SP-A1; SP-A1 beta; SP-A1 delta; SP-A1 epsilon; SP-A1 gamma; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant associated protein A; surfactant associated protein A1; surfactant protein A1; surfactant protein A1 variant AB'D' 6A; surfactant protein A1 variant AB'D' 6A2; surfactant protein A1 variant AB'D' 6A3; surfactant protein A1 variant AB'D' 6A4; surfactant protein A1 variant ACD' 6A; surfactant protein A1 variant ACD' 6A2; surfactant protein A1 variant ACD' 6A3; surfactant protein A1 variant ACD' 6A4; surfactant protein A1 variant AD' 6A; surfactant protein A1 variant AD' 6A2; surfactant protein A1 variant AD' 6A3; surfactant protein A1 variant AD' 6A4; surfactant protein A1B; surfactant protein A2; surfactant pulmonary associated protein A1; surfactant, pulmonary-associated protein A1; surfactant, pulmonary-associated protein A1A; surfactant, pulmonary-associated protein A1B; surfactant, pulmonary-associated protein A2A; Surfactant-associated protein 1 (pulmonary surfactant protein SP-A); Surfactant-associated protein 1 (pulmonary surfactant protein, SP-A) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20387, 24773, 653509, 729238 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P08427, P35242, Q8IWL1, Q8IWL2 | |
| Sftpa1, SFTPA2 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human SFTPA1/2 (206-237aa VNYTNWYRGEPAGRGKEQCVEMYTDGQWNDRN). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction