Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116173
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116173 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116173 Supplier Invitrogen™ Supplier No. PA5116173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SFTPA2 Polyclonal Antibody for Western Blot

This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPA2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SFTPA2
Gene Accession No. Q8IWL1
Gene Alias 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC5; collectin 5; Collectin-5; PSAP; PSPA; PSP-A; Pulmonary surfactant-associated protein A2; SFTP1; SFTPA; SFTPA2; SFTPA2B; SP-2A; SP-2A beta; SP-2A gamma; SP-A; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant protein A2; surfactant, pulmonary-associated protein A2A
Gene Symbols SFTPA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 174-202 amino acids from the C-terminal region of human SFTPA2B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729238
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.