Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFTPB Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ SA172016
Description
SA1-72016 detects Surfactant Protein B in human, mouse, and rat samples. SA1-72016 has been successfully used in western blot procedures. The SA1-72016 immunogen is from Chicken immunized with SP-B purified from Infasurf.
Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
Specifications
SFTPB | |
Polyclonal | |
Unconjugated | |
SFTPB | |
18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B | |
Chicken | |
IgY fraction | |
RUO | |
507398 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
20 mg/mL | |
PBS with 0.075% sodium azide; pH 7.2 | |
P15781 | |
SFTPB | |
SP-B purified from Infasurf. | |
10 mg | |
Primary | |
Bovine | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction