Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPB Precursor Monoclonal Antibody (SPB01 (1-2-A5-9)), Invitrogen™

Catalog No. PIMA513975
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513975 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513975 Supplier Thermo Scientific Supplier No. MA513975
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13975 targets Surfactant Protein B (Precursor) in IF and IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-13975 immunogen is recombinant pro-surfactant protein B.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPB Precursor
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPB01 (1-2-A5-9)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene SFTPB
Gene Accession No. P07988, P22355
Gene Alias 18 kDa pulmonary-surfactant protein; 6 kDa protein; PSP-B; Pulmonary surfactant-associated protein B; pulmonary surfactant-associated proteolipid SPL(Phe); SFTB3; SFTP3; SFTPB; SMDP1; SP-B; surfactant associated protein B; surfactant protein B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B
Gene Symbols SFTPB
Host Species Mouse
Immunogen Recombinant pro-surfactant protein B
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192155, 6439
Target Species Human, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.