Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPC Mouse anti-Human, Clone:5E6A9, Abnova™

Catalog No. 89334611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-611 Supplier Abnova Corporation Supplier No. MAB17756
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human SFTPC.

The SFTPC gene encodes pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. It is produced exclusively by type II alveolar epithelial cells in the lung. Pulmonary surfactant is a lipid-rich material that prevents lung collapse by lowering surface tension at the air-liquid interface in the alveoli of lung. Surfactant is composed of phospholipids and other surfactant-associated proteins (Clark and Clark, 2005 [PubMed 15927881]). See also SFTPA1 (MIM 178630), SFTPB (MIM 178640), and SFTPD (MIM 178635).[supplied by OMIM]

Specifications

Antigen SFTPC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E6A9
Conjugate Unconjugated
Description surfactant protein C
Dilution ELISA (1:10000)
Flow Cytometry
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SFTPC
Gene Alias PSP-C, SFTP2, SMDP2, SP-C
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 60-180 of human SFTPC from E. coli.
Quantity 100 μg
Gene ID (Entrez) 6440
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.