Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579989
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579989 Supplier Invitrogen™ Supplier No. PA579989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Lung Tissue, Rat Brain Tissue, PANC whole cell. IHC: human lung cancer tissue IHC-F: human placenta tissue. ICC/IF: A549 Cell, Hela Cell.

Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPD
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene SFTPD
Gene Accession No. P35247, P35248
Gene Alias AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Gene Symbols SFTPD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Surfactant protein D (292-321aa RSAAENAALQQLVVAKNEAAFLSMTD SKTE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25350, 6441
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.