Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPD Recombinant Rabbit Monoclonal Antibody (001)
GREENER_CHOICE

Catalog No. PIMA529847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529847 Supplier Invitrogen™ Supplier No. MA529847
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rhesus SFTPD/SP-D/SFTP4.

Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPD
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SFTPD
Gene Accession No. Q1PBC5
Gene Alias AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Gene Symbols SFTPD
Host Species Rabbit
Immunogen Recombinant Rhesus SFTPD/SP-D/SFTP4 protein (Met1-Phe375).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 678657
Target Species Rhesus Macaque
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.