Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP2411530.025MG
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA.Specifications
SFTS Virus HB29 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot, ELISA 1 μg/mL | |
B303_sMgp1, Membrane glycoprotein polyprotein | |
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 60 - 110 of SFTS Virus HB29. Amino Acid Squence: QRGGRSQVSYYPAENSYSR | |
0.025 mg | |
Primary | |
Virus | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction