Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | SFTS Virus HB29 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for ELISA.Specifications
SFTS Virus HB29 | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against an 18 amino acid synthetic peptide near the center of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 510 - 560 of SFTS Virus HB29. Amino Acid Squence: HEEIGENGEGNQDDVRIE | |
Primary | |
SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Virus | |
B303_sMgp1, Membrane glycoprotein polyprotein | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title