Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SFX-Insect™ Cell Culture Media

Catalog No. SH3027801
Click to view available options
Quantity:
500 mL
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188
Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Phenol Red, No HEPES, No Protein
No Protein, No HEPES
No Protein, No Phenol Red, No HEPES
No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES
Form:
Liquid
Powder
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.

  • Protein free media
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Insect Cell Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188
Without Additives No Phenol Red, No HEPES, No Protein
Quantity 500 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.