Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFXN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588887 Supplier Invitrogen™ Supplier No. PA588887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MILIGRMSAQ VPMNMTITGC MMTFYRTTPA VLFWQWINQS FNAVVNYTNR SGDAPLTVNE LGTAYVSATT GAVATALGLN ALTKHVSPLI GRFVPFAAVA AANCINIPLM RQRELKVGIP VTDENGNRLG ESANAAKQAI TQVVVSRILM AAPGMAIPPF IMNTLEKKAF LKRFPWMSAP IQVGLVGFCL VFATPLCCAL FPQKSSMSVT SLEAELQAKI QESHPELRRV YFNKGL; Positive Samples: MCF-7, 22RV1, LO2, HeLa, Mouse liver, Mouse brain, Rat liver; Cellular Location: Mitochondrion membrane, Multi-pass membrane protein.

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFXN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SFXN1
Gene Accession No. Q63965, Q99JR1, Q9H9B4
Gene Alias 2810002O05Rik; A930015P12Rik; f; flexed tail; flexed-tail; sfxn1; sideroflexin 1; sideroflexin-1; TCC; tricarboxylate carrier protein; zgc:100851
Gene Symbols SFXN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 87-322 of human SFXN1 (NP_0735912).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14057, 364678, 94081
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.