Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFXN1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA584617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584617 Supplier Thermo Scientific Supplier No. PA584617

Rabbit Polyclonal Antibody

Immunogen sequence: SSMSVTSLEA ELQAKIQESH PELRRVYFNK GL

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFXN1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SFXN1
Gene Accession No. Q9H9B4
Gene Alias 2810002O05Rik; A930015P12Rik; f; flexed tail; flexed-tail; sfxn1; sideroflexin 1; sideroflexin-1; TCC; tricarboxylate carrier protein; zgc:100851
Gene Symbols SFXN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SFXN1
Purification Method Anitgen affinity chromatography, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.