Learn More
SFXN1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305443A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Sheep, Bovine and Pig
SFXN1 might be involved in the transport of a component required for iron utilization into or out of the mitochondria.Specifications
SFXN1 | |
Polyclonal | |
Unconjugated | |
SFXN1 | |
TCC, sideroflexin-1, tricarboxylate carrier protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94081 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9H9B4 | |
SFXN1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.