Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFXN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120586
This item is not returnable.
View return policy
Description
Positive test controls include: HepG2, Mouse kidney, Rat kidney, Rat pancreas. Immunogen sequence: IDAPRWDQRT FLGRVKHFLN ITDPRTVFVS ERELDWAKVM VEKSRMGVVP PGTQVEQLLY AKKLYDSAFH PDTGEKMNVI G.
Specifications
| SFXN2 | |
| Polyclonal | |
| Unconjugated | |
| SFXN2 | |
| C79732; F630107H02Rik; Sfxn2; sideroflexin 2; sideroflexin-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 118980, 294011, 94279 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.29 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q925N2, Q96NB2 | |
| SFXN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-90 of human SFXN2 (NP_849189.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction