Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFXN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552505 Supplier Invitrogen™ Supplier No. PA552505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESKMGELPLD INIQEPRWDQ STFLGRARHF FTVTDPRNLL LSGAQLEASR NIVQNYRAGV VTPGITEDQL WRAKYVYDSA FHPDTGEKVV LIGRMSAQVP MN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

Mitochondrial serine transporter that mediates transport of serine into mitochondria, an important step of the one-carbon metabolism pathway (PubMed:30442778). Mitochondrial serine is converted to glycine and formate, which then exits to the cytosol where it is used to generate the charged folates that serve as one-carbon donors. Also catalyzes an electroneutral exchange of tricarboxylate, dicarboxylate, or phosphoenolpyruvate across the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFXN3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SFXN3
Gene Accession No. Q91V61, Q9BWM7, Q9JHY2
Gene Alias BA108L7.2; SFX3; SFXN3; sideroflexin 3; sideroflexin-3; TCC; tricarboxylate carrier; tricarboxylate carrier-like protein
Gene Symbols SFXN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SFXN3. Recombinant protein control fragment (Product #RP-90095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65042, 81855, 94280
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.