Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFXN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535980
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Potential iron transporter.
Specifications
SFXN4 | |
Polyclonal | |
Unconjugated | |
SFXN4 | |
BCRM1; Breast cancer resistance marker 1; COXPD18; SFXN4; sideroflexin 4; sideroflexin-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
119559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6P4A7 | |
SFXN4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 201 of SFXN4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction