Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SG2NA Antibody (S68), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB11074572
Description
SG2NA Monoclonal specifically detects SG2NA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| SG2NA | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.02% Sodium Azide | |
| Cell cycle autoantigen SG2NA, cell cycle S/G2 nuclear autoantigen, GS2NA, S/G2 antigen, SG2NAnuclear autoantigen, striatin, calmodulin binding protein 3, striatin-3 | |
| Mouse | |
| 90 kDa | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 29966 | |
| Human, Mouse, Rat | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| S68 | |
| Western Blot 1:250-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500 | |
| Q13033 | |
| STRN3 | |
| Synthetic peptide made to an internal portion of human SG2NA (within residues 200-300). [Swiss-Prot# Q13033] | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody is specific for SG2NA. It does not cross-react with Striatin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction