Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SG2NA Antibody (S68), Novus Biologicals™
SDP

Catalog No. NB11074572T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11074572T 0.025 mL
NB11074572 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11074572T Supplier Novus Biologicals Supplier No. NB11074572SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

SG2NA Monoclonal specifically detects SG2NA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen SG2NA
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone S68
Conjugate Unconjugated
Dilution Western Blot 1:250-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500
Formulation Ascites with 0.02% Sodium Azide
Gene Accession No. Q13033
Gene Alias Cell cycle autoantigen SG2NA, cell cycle S/G2 nuclear autoantigen, GS2NA, S/G2 antigen, SG2NAnuclear autoantigen, striatin, calmodulin binding protein 3, striatin-3
Gene Symbols STRN3
Host Species Mouse
Immunogen Synthetic peptide made to an internal portion of human SG2NA (within residues 200-300). [Swiss-Prot# Q13033]
Molecular Weight of Antigen 90 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29966
Test Specificity This antibody is specific for SG2NA. It does not cross-react with Striatin.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.