Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SGC 707
GREENER_CHOICESDP

Catalog No. 536750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-675-0 50 mg
53-671-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-675-0 Supplier R&D Systems™ Supplier No. 5367/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective allosteric inhibitor of PRMT3

SGC 707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 31 nM). Inhibits PRMT3-mediated methyltransferase activity in vitro. Displays >100-fold selectivity against 31 other methyltransferases and 250 other non-epigenetic targets. Decreases H4R3me2a levels in HEK293 cells in vitro.

Negative control also available.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type SGC 707
Purity >98%
Molecular Weight (g/mol) 298.34
Inhibitors SGC 707

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.