Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SGC-CBP30

Catalog No. 488950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-895-0 50 mg
48-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-895-0 Supplier Tocris Bioscience™ Supplier No. 4889/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent CBP/p300 BRD inhibitor

Potent CBP/p300 bromodomain (BRD) inhibitor (Kd values are 21 and 32 nM for CBP and p300 BRDs respectively). Exhibits 40-fold and 250-fold selectivity for CBP over the first BRD of BRD4 (BRD4(1)) and BRD4(2) respectively. Accelerates FRAP recovery in cells at a concentration of 1 μM.

Chemical Identifiers

CAS 1613695-14-9
Molecular Formula C28H33ClN4O3

Specifications

Quantity 50 mg
Formula Weight 509.04
Purity >98%
Molecular Weight (g/mol) 509.05
Chemical Name or Material SGC-CBP30
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.