Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SGC SMARCA-BRDVIII
GREENER_CHOICESDP

Catalog No. 746050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-605-0 50 mg
74-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-605-0 Supplier Tocris Bioscience™ Supplier No. 7460/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective SMARCA2/4 and PB1(bromo 5)-selective SWI/SNF bromodomain inhibitor

SGC-SMARCA-BRDVIII is a selective and high affinity SMARCA2/4 and PB1(bromo 5)-selective SWI/SNF bromodomains (Kd(ITC) values are 35, 36 and 13 nM, respectively). SGC-SMARCA-BRDVIII exhibits selectivity over 25 other bromodomain families and shows no off-target activity on 85 protein kinases screened using temperature-shift binding assays. SGC-SMARCA-BRDVIII suppresses formation of 3T3-L1 mouse fibroblasts into adipocytes (EC50 <1.0 μM).

Specifications

Chemical Name or Material 1,1-Dimethylethyl 4-[3-amino-6-(2-hydroxyphenyl)-4-pyridazinyl]-1-piperazinecarboxylate bromodomains
CAS 1997319-84-2
Quantity 50 mg
Target Bromodomain Inhibitors
Molecular Formula C19H25N5O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.