Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552835 Supplier Invitrogen™ Supplier No. PA552835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FHMGGNMELK AENSIILNGS VMVSTTRLPS SSSGDQLGSG DWVRYKLCMC ADGTLFKVQV TSQNMGCQIS DNPCGNTH Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGCB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGCB
Gene Accession No. Q16585
Gene Alias 43 kDa dystrophin-associated glycoprotein; 43DAG; 43kDa; A3b; AI747103; AI844814; beta sarcoglycan; beta-sarcoglycan; beta-sarcoglycan(43kD dystrophin-associated glycoprotein); beta-SG; fa03a05; LGMD2E; limb girdle muscular dystrophy 2E (non-linked families); sarcoglycan beta; sarcoglycan, beta (43kD dystrophin-associated glycoprotein); sarcoglycan, beta (43kDa dystrophin-associated glycoprotein); sarcoglycan, beta (dystrophin-associated glycoprotein); SGC; Sgcb; wu:fa03a05; zgc:110833
Gene Symbols SGCB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGCB. Recombinant protein control fragment (Product #RP-90379).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.