Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGCD Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142694
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142694 Supplier Invitrogen™ Supplier No. PA5142694
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 (NP_000328.2) and isoform 3 (NP_001121681.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGCD
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SGCD
Gene Accession No. Q92629
Gene Alias 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kDa; CMD1L; CMD1LSG-delta; DAGD; delta-sarcoglycan; Delta-SG; dystrophin associated glycoprotein, delta sarcoglycan; LGMD2F; MGC22567; placental delta sarcoglycan; sarcoglycan delta; sarcoglycan, delta (35kD dystrophin-associated glycoprotein); sarcoglycan, delta (35kDa dystrophin-associated glycoprotein); sarcoglycan, delta (dystrophin-associated glycoprotein); sarcoglycandelta; sarcoglycan-delta; SGCD; SGCDP; SGD; SGdelta; SG-delta
Gene Symbols SGCD
Host Species Goat
Immunogen Peptide with sequence C-RLPHGSYTPTGTR (aa 244-256).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497892, 6444
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.