Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567363 Supplier Invitrogen™ Supplier No. PA567363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIQLVHHSAIQ KSTKELRDMS KNREIAWPLS TLPVFHPVTG EIIPPLHTDN YDSTNMPLMQ TQQNLPHQTQ IPQQQTTGKW Y Highest antigen sequence identity to the following orthologs - mouse 94%, rat 96%.

This gene encodes the epsilon member of the sarcoglycan family. Sarcoglycans are transmembrane proteins that are components of the dystrophin-glycoprotein complex, which link the actin cytoskeleton to the extracellular matrix. Unlike other family members which are predominantly expressed in striated muscle, the epsilon sarcoglycan is more broadly expressed. Mutations in this gene are associated with myoclonus-dystonia syndrome. This gene is imprinted, with preferential expression from the paternal allele. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGCE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGCE
Gene Accession No. O43556
Gene Alias dystonia 11, myoclonic; DYT11; EMBL:AAO37579.1, ECO:0000312; EMBL:AAO37579.1}; Epsilon sarcoglycan; Epsilon SG; Epsilon-sarcoglycan; epsilon-sarcoglycan {ECO:0000312; epsilon-SG; epsilon-SG {ECO:0000250; ESG; e-SG; RGD:1303201}; sarcoglycan; sarcoglycan epsilon; sarcoglycan, epsilon; SGCE; sgce {ECO:0000312; UniProtKB:O43556}; UNQ433/PRO840
Gene Symbols SGCE
Host Species Rabbit
Immunogen Recombinant Human SGCE. Recombinant protein control fragment (Product #RP-105591).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8910
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.