Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGCG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587970 Supplier Invitrogen™ Supplier No. PA587970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVTKDGLRLE GESEFLFPLY AKEIHSRVDS SLLLQSTQNV TVNARNSEGE VTGRLKVGPK MVEVQNQQFQ INSNDGKPLF TVDEKEVVVG TDKLRVTGPE GALFEHSVET PLVRADPFQD LRLESPTRSL SMDAPRGVHI QAHAGKIEAL SQMDILFHSS DGMLVLDAET VCLPKLVQGT WGPSGSSQSL YEICVCPDGK LYLSVAGVST TCQEHSHICL; Positive Samples: HeLa, MCF-7; Cellular Location: Cell membrane, Cytoplasm, Single-pass type II membrane protein, cytoskeleton, sarcolemma.

Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGCG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Sgcg
Gene Accession No. Q13326
Gene Alias 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kD dystrophin-associated glycoprotein; 35kDa; 5430420E18Rik; A4; AI642964; DAGA4; DMDA; DMDA1; gamma sarcoglycan; gamma-sarcoglycan; gamma-SG; LGMD2C; MAM; sarcoglycan gamma; sarcoglycan, gamma; sarcoglycan, gamma (35kD dystrophin-associated glycoprotein); sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein); sarcoglycan, gamma (dystrophin-associated glycoprotein); SCARMD2; SCG3; SGCG; sgcg gene for gamma-sarcoglycan; TYPE; zgc:100924
Gene Symbols Sgcg
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 72-291 of human SGCG (NP_0002221).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6445
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.