Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 181021AP150UL

Description
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.Specifications
SGCG | |
Polyclonal | |
Unconjugated | |
SGCG | |
35DAG, A4, DAGA4, DMDA, DMDA1, Gamma sarcoglycan, Gamma SG, LGMD2C, MAM, SCARMD2, SCG3, SGCG, TYPE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24053, 305941, 6445 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P82348, Q13326 | |
SGCG | |
SGCG Fusion Protein Ag12402 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction