Learn More
Invitrogen™ SGCZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110365
Description
Immunogen sequence: NLRVTKKGIR LEGISEFLLP LYVKEIHSRK DSPLVLQSDR NVTVNARNHM GQLTGQLTIG ADAVEAQCKR FEVRASEDGR VLFSADEDEI TIGAEKLKVT GTEGAVFGHS VETPHIRAEP SQDLRLESPT RSLIMEAPRG VQVSAAAGDF KATCRKELHL QSTEGEIFLN AETIKLGNLP TGSFSSSSPS SSSSRQTVYE LCVCPNGKLY LSPAGVGSTC QSSSNICLWS.
The zeta-sarcoglycan gene measures over 465 kb and localizes to 8p22. This protein is part of the sarcoglycan complex, a group of 6 proteins. The sarcoglycans are all N-glycosylated transmembrane proteins with a short intra-cellular domain, a single transmembrane region and a large extra-cellular domain containing a carboxyl-terminal cluster with several conserved cysteine residues. The sarcoglycan complex is part of the dystrophin-associated glycoprotein complex (DGC), which bridges the inner cytoskeleton and the extra-cellular matrix.
Specifications
| SGCZ | |
| Polyclonal | |
| Unconjugated | |
| SGCZ | |
| C230085N17Rik; sarcoglycan zeta; sarcoglycan, zeta; Sgcz; Zeta-sarcoglycan; zeta-SG; ZSG1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 137868, 244431, 364605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8BX51, Q96LD1 | |
| SGCZ | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 83-312 of human SGCZ (NP_631906.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.