Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGCZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553610 Supplier Invitrogen™ Supplier No. PA553610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLYVKEIHSR KDSPLVLQSD RNVTVNARNH MGQLTGQLTI GADAVEAQCK RFEVRASEDG RVLFSADEDE ITIGAEKLKV TGTEGAVFGH SVETPHIRAE PSQDL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 57%.

The zeta-sarcoglycan gene measures over 465 kb and localizes to 8p22. This protein is part of the sarcoglycan complex, a group of 6 proteins. The sarcoglycans are all N-glycosylated transmembrane proteins with a short intra-cellular domain, a single transmembrane region and a large extra-cellular domain containing a carboxyl-terminal cluster with several conserved cysteine residues. The sarcoglycan complex is part of the dystrophin-associated glycoprotein complex (DGC), which bridges the inner cytoskeleton and the extra-cellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGCZ
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGCZ
Gene Accession No. Q96LD1
Gene Alias C230085N17Rik; sarcoglycan zeta; sarcoglycan, zeta; Sgcz; Zeta-sarcoglycan; zeta-SG; ZSG1
Gene Symbols SGCZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGCZ. Recombinant protein control fragment (Product #RP-90548).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.