Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Specifications
| Type | Ferrule |
| Packaging Quantity | 10 ferrules, 2 nuts |
| For Use With | 0.10 to 0.25 mm Column |
| Material | SilTite |