Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ BPX Dioxin Capillary Columns
SDP

Catalog No. SG054235 Shop All SGE Inc. Products
Change view
Click to view available options
Length (Metric):
60 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
SG-054235 60 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SG-054235 Supplier Trajan™ Supplier No. 054235

missing translation for 'validMainframeMsg'

SGE BPX Dioxin GC Capillary Columns arespecifically designed for analysis or dioxins and related compounds

  • High temperature, low bleed column
  • Non-polar
  • Inert

Specifications

Diameter (Metric) Inner 0.25 mm
Film Thickness 0.25 μm
Length (Metric) 60 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.