Learn More
Trajan™ Methyl Deactivated Fused Silica Capillary Tubing
Essential for maintaining a high performance chromatography system.Trajan Scientific and Medical™ Methyl Deactivated Fused Silica Capillary Tubing is chemically inert and thermally stable up to 380°C.
Supplier: Trajan™ 064034
Description
- Suitable for organic and aqueous solvents
- Methyl deactivated for use with hydrocarbons (pentane, hexane, heptane, iso-octane, aromatics and mixed solvents - n-paraffins/chlorinated solvents
- Individually tested performance of each 30m length from which all shorter lengths are prepared
- Ideal for biotechnology applications
Specifications
Capillary Tubing | |
0.020 in. | |
0.53 mm | |
82 ft. | |
Fused Silica | |
380°C |
Ideal for Biotechnology AppliCatIons, and Organic and Aqueous Solvents | |
0.026 in. | |
0.68 mm | |
25 m | |
716°F |