Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
SDP
Catalog No. SG002760 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
10 μL
Model:
10R-AG-GT-MS1
Catalog No. Volume (Metric) Model
SG002760 10 μL 10R-AG-GT-MS1
1 options
Catalog No. SG002760 Supplier Trajan™ Supplier No. 002760
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
For Use With (Application) Improves mass spectrometry analysis sensitivity
Product Type Diamond MS Syringe
Model 10R-AG-GT-MS1
Quantity Ea.
Volume (Metric) 10 μL
Connection Type Removable